Benzamidine hydrochloride hydrate ≥98% (by anhydrous basis), pulver, Ultra Pure Grade

Leverandør: VWR Chemicals
Warning

0616-100GEA 0 NOK
0616-100G 0616-25G
Benzamidine hydrochloride hydrate ≥98% (by anhydrous basis), pulver, Ultra Pure Grade
Benzamidine hydrochloride hydrate

A protease inhibitor of arginine-selective enzymes like trypsin, kallikrein and thrombin.


  • Fine white crystalline powder

Formel: C₆H₅C(=NH)NH₂·HCl·H₂O
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00066285
CAS-nummer: 206752-36-5
EINECS: 216-795-4

Bestill nå

Spesifikasjon Testresultater

Em (lambda max, MEOH, anhy) ≥9500
Heavy Metals (as Pb) ≤0.005%
Melting point 77 – 87 °C
Purity (anhydrous) ≥98%
Solubility (1%, water) Pass

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR