Sitronsyre monohydrat 99.5-100.5% (by anhydrous basis), granular USP, Macron Fine Chemicals™

Leverandør: Avantor Performance Materials
Warning

0616-06EA 2750 NOK
0616-06 0616-88
Sitronsyre monohydrat 99.5-100.5% (by anhydrous basis), granular USP, Macron Fine Chemicals™
Sitronsyre monohydrat

NB: Must be subjected to further processing during the preparation of injectable dosage forms.

Formel: C₆H₈O₇.H₂O
Smeltepunkt: 135…152 °C
Tetthet: 1,552 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00149972
CAS-nummer: 5949-29-1
EINECS: 201-069-1
Merck Indeks: 13,02350
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3160681f96abd248f56672a96b72cc5f6e9d89a2&vid=c4db52c32c630d2c9876e60fc3e9e70a117ea0d8&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Meets U.S.P Requirements
GMP Manufactured Product
Preserve in Tight Containers
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical
USP - Assay (as C₆H₈O₇) (calculated on anhydrous basis) 99.5 - 100.5 %
USP - Clarity of Solution Passes Test
USP - Color of Solution Passes Test
USP - Identification Passes Test
USP - Water (H₂O)(by Karl Fischer titrn) 7.5 - 9.0 %
USP - Residue on Ignition ≤0.1 %
USP - Readily Carbonizable Substances Passes Test
USP - Sulfate (SO₄) ≤0.015 %
USP - Limit of Oxalic Acid ≤0.036 %
Solubility (5 in 50) Passes Test
No Class 1,2,3 or other solvents are used or produced in the
manufacturing or purification of the product.
Must be subjected to further processing during the preparation of
injectable dosage forms.
Elemental Impurities (USP 232, EP 5.20) - Information on elemental
impurities for this product is available on the associated Product
Regulatory Data Sheet and elemental impurity profile report.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR