Phenol red sodium salt ACS

Leverandør: VWR Chemicals

Synonymer: 3,3-Bis(4-hydroxyphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide, sodium salt, Phenol red water soluble, Sodium hydrogen 4,4'-(3H-2,1-benzoxathiol-3-ylidene)bisphenolate S,S-dioxide

0648-50GEA 2675 NOK
0648-50G
Phenol red sodium salt ACS
Phenol red sodium salt
Formel: C₁₉H₁₃NaO₅S
MW: 376.35 g/mol
Smeltepunkt: 285
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00066901
CAS-nummer: 34487-61-1
EINECS: 252-057-8
Merck Indeks: 12,07397

Bestill nå

Spesifikasjon Testresultater

Solubility (0.1%, water) Pass
Visual transition interval (pH 6.8 – 8.2) Pass

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR