Toluidinblått O, high purity

Leverandør: VWR Chemicals

0672-25GEA 7450 NOK
0672-25G 0672-50G
Toluidinblått O, high purity
Toluidinblått O

A blue counterstain often used for metachromatic staining of mast cells, cartilage, and acid mucins.


  • Black-grey powder

Formel: C₁₅H₁₆ClN₃S
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00011934
CAS-nummer: 92-31-9
EINECS: 202-146-2
Merck Indeks: 12,09658

Bestill nå

Spesifikasjon Testresultater

Dye Content >50.0%
Em (626nm, Water)
Loss on Drying ≤8.0%
Color Index 52040


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR