Supelco® Organic Reference Standard, (+)-Valencene

Leverandør: Merck

06808-50MG
06808-50MGEA 2950 NOK
06808-50MG
Supelco® Organic Reference Standard, (+)-Valencene
Standarder Organiske standarder

NB: P301 + P310 - P331 - P405 - P501

https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c8a662238c6969b6f9d3eafd9e4aaa2a4d5087eb&vid=c8a662238c6969b6f9d3eafd9e4aaa2a4d5087eb&prid=web&clid=SAPDAM

Bestill nå

SPESIFIKASJONER

Synonym(s) (3R,4aS,5R)-4a,5-Dimethyl-3-isopropenyl-1,2,3,4,4a,5,6,7-octahydronaphthalene
M-Clarity quality level MQ100
Environmentally Preferable
Tetthet 0,92 g/ml at 25 °C (lit.)
CAS-nr 4630-07-3
MDL No. MFCD00075884
Formel C₁₅H₂₄
Molekylvekt 204,35
kokepunkt 274 °C (lit.)
Hardhetsklasse beskrivelse H304
Flammepunkt 100 °C - closed cup
Fraktforhold STD
Lagringsforhold 2...8 °C
Lagringstemp 2…8 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR