Organic reference standard, 4-Deoxy-L-erythronic acid sodium salt hydrate, ≥97,0% (GC), Supelco®
Leverandør: Merck
06932-50MG
06932-50MGEA 5375 NOK
06932-50MG
Organic reference standard, 4-Deoxy-L-erythronic acid sodium salt hydrate, ≥97,0% (GC), Supelco®
Standarder Organiske standarder
4-Deoxyerythronic acid has been associated with uremia and diabetes mellitus type 1, but is also a normal organic acid present in human biofluids, derived presumably from L-threonine.