Organic reference standard, Tris(2-chloroethyl) phosphate, analytical standard, Supelco®
Leverandør: Merck
07296-1ML
07296-1MLEA 830 NOK
07296-1ML
Organic reference standard, Tris(2-chloroethyl) phosphate, analytical standard, Supelco®
Standarder Organiske standarder
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c3d1bb65368d107017f4339dcacb537b140489e3&vid=c3d1bb65368d107017f4339dcacb537b140489e3&prid=web&clid=SAPDAM
Bestill nå
SPESIFIKASJONER
Beskrivelse Tris(2-chloroethyl) phosphate
Environmentally Preferable
CAS-nr 115-96-8
Formel (ClCH₂CH₂O)₃PO
kokepunkt 192 °C/10 mm Hg (lit.)