Diammoniumhydrogenortofosfat 98,0 Acidimetry, BAKER ANALYZED® ACS, J.T.Baker®

Leverandør: Avantor
Warning

Synonymer: Diammonium hydrogen orthophosphate, Ammonium hydrogen phosphate, di-Ammonium hydrogen phosphate, Ammonium Phosphate, Ammonium biphosphate, Fyrex, Ammonium phosphate dibasic, Ammonium monohydrogen phosphate, Diammonium hydrogen phosphate

0784-01EA 0 NOK
0784-01 0784-07 0784-R
Diammoniumhydrogenortofosfat 98,0 Acidimetry, BAKER ANALYZED® ACS, J.T.Baker®
Diammoniumhydrogenortofosfat

Crystals. Lot analysis on label.

Formel: (NH₄)₂HPO₄
MW: 132.06 g/mol
Smeltepunkt: 185
Tetthet: 1.62 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00010891
CAS-nummer: 7783-28-0
EINECS: 231-987-8
REACH: 01-2119490974-22
Merck Indeks: 13,00546

Bestill nå

Spesifikasjon Testresultater

Meets ACS Reagent Chemical Requirements ≥98.0%
For Laboratory,Research,or Manufacturing Use 7.7 - 8.1
Assay ((NH₄)₂HPO₄) (by acidimetry) ≥98.0 %
pH of 5% Solution at 25°C 7.7 - 8.1
Insoluble Matter ≤0.005 %
Chloride (Cl) ≤0.001 %
Nitrate (NO₃) ≤0.003 %
Sulfate (SO₄) ≤0.01 %
Heavy Metals (as Pb) ≤0.001 %
Trace Impurities - Iron (Fe) ≤0.001 %
Calcium (Ca) ≤0.001 %
Magnesium (Mg) ≤0.0005 %
Potassium (K) ≤0.005 %
Sodium (Na) ≤0.005 %
Meets Reagent Specifications for testing USP/NF monographs


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR