Sudan II, Supelco®

Leverandør: Merck

Synonymer: 1-(2,4-Dimethylphenylazo)-2-naphthol, Solvent Orange 7, 1-((2,4-Dimethylphenyl)diazenyl)naphthalen-2-ol

07937-25MG
07937-25MGEA 1650 NOK
07937-25MG
Sudan II, Supelco®
Sudan II
Formel: C₁₈H₁₆N₂O
MW: 276.34 g/mol
MDL nummer: MFCD00003896
CAS-nummer: 3118-97-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8ddd43aa1bb60a68b4b6b5a8743e0b397944a82f&vid=fdc306d0259efb0d6b385abe7fabd6d97a61c961&prid=web&clid=SAPDAM

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR