Guanidiniumklorid, GenAR® for biotechnology, Macron Fine Chemicals™

Leverandør: Avantor

Synonymer: Guanadine hydrochloride, Aminoformamidine hydrochloride, Aminomethanamidine hydrochloride, Guanidine HCI, Guanidine chloride, Guanidinium chloride, Guanidine hydrochloride

0834-26EA 0 NOK
0834-26
Guanidiniumklorid, GenAR® for biotechnology, Macron Fine Chemicals™
Guanidiniumklorid
Formel: NH₂C(=NH)NH₂·HCl
MW: 95.53 g/mol
Smeltepunkt: 185
MDL nummer: MFCD00013026
CAS-nummer: 50-01-1
Merck Indeks: 13,04578
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=da5a871cab977e6d9dc666b457732eb1e869a714&vid=b0d4fcb044b6106fce8e40448dbf2ce6e7077ad7&prid=web&clid=SAPDAM

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR