Natriumhydrogenkarbonat 99.7-100.3% ACS

Leverandør: VWR Chemicals

0865-1KGEA 2025 NOK
0865-1KG 0865-2.5KG
Natriumhydrogenkarbonat 99.7-100.3% ACS
Natriumhydrogenkarbonat
Formel: NaHCO₃
Smeltepunkt: 270 °C
Tetthet: 2,159 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00003528
CAS-nummer: 144-55-8
EINECS: 205-633-8
Merck Indeks: 13,08655
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f6b1b80d2afb3deab0c4ca836a921fcaad637890&vid=1163b37d9cf1c10fe0ed252e02a51fe12d6b50dd&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Ammonium <0.0005%
Calcium ≤0.02%
Chloride ≤0.003%
Heavy metals (as Pb) ≤0.0005%
Insolubles ≤0.015%
Iron ≤0.001%
Magnesium ≤0.005%
Phosphate ≤0.001%
Potassium ≤0.005%
Purity 99.7 - 100.3%
Sulphur compounds ≤0.003%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR