Fenol rødt, powder ACS pH-indicator

Leverandør: VWR Chemicals

Synonymer: 3,3-Bis(4-hydroxyphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide

0882-25GEA 1950 NOK
0882-25G
Fenol rødt, powder ACS pH-indicator
Fenol rødt

Bright red powder

Formel: C₁₉H₁₄O₅S
MW: 354.38 g/mol
Smeltepunkt: > 300 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00003552
CAS-nummer: 143-74-8
EINECS: 205-609-7
Merck Indeks: 13,07329

Bestill nå

Spesifikasjon Testresultater

Solubility (0.1%, Alcohol) Pass
Visual Transition Interval (pH 6.8 - 8.2) Pass


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR