Dextrose solution 50% (w/w), Biotech Reagent, J.T.Baker®

Leverandør: Avantor

Synonymer: Corn sugar, ᴅ-(+)-Glucose, Glucose, D-(+)-Dextrose, (2R,3S,4R,5R)-2,3,4,5,6-Pentahydroxyhexanal, Glucolin, Dextrose, D-Glucose

Management of Change (MOC) category = R

0890-09EA 0 NOK
0890-09
Dextrose solution 50% (w/w), Biotech Reagent, J.T.Baker®
D(+)-Glukose
Formel: C₆H₁₂O₆
MW: 180.16 g/mol
Smeltepunkt: 146
MDL nummer: MFCD00148912
CAS-nummer: 50-99-7
Merck Indeks: 14,04459

Bestill nå

Spesifikasjon Testresultater

GMP Manufactured Product
CAUTION: For Manufacturing, processing or repackaging
Assay (w/w) 49 - 51 %
Osmolality, mOsm/kg 317 - 393
Endotoxin Concentration (EU/g) ≤1
No Class 1,2,3 or other solvents are used or produced in the manufacturing or purification of the product.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR