iso-Amylalkohol ≥99%, Ultra Pure Grade

Leverandør: VWR Chemicals
Danger

Synonymer: Isopentyl alcohol, iso-Pentanol, 3-Methyl-1-butanol, 3-Methylbutanol

0944-1LEA 3100 NOK
0944-1L
iso-Amylalkohol ≥99%, Ultra Pure Grade
iso-Amylalkohol
Formel: (CH₃)₂CHCH₂CH₂OH
MW: 88.15 g/mol
Kokepunkt: 130
Smeltepunkt: –117 °C
Tetthet: 0.812 g/cm³ (20 °C)
Flash punkt: 42
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00002934
CAS-nummer: 123-51-3
EINECS: 204-633-5
FN: 1105
ADR: 3 III
Merck Indeks: 13,05214
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cd7b89cc1890b5d4b2083195c30c0d1047c32e9a&vid=170c60e26c7b4edab6e0686f91d04e676da16e6d&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Color (APHA) <20
Moisture (KF) ≤1%
Purity ≥99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR