Ammoniumjern(III)citrat (14,5 - 16,0% Fe) (RT), green, powder, Purum p.a.

Leverandør: Honeywell Chemicals

Synonymer: Ammonium ferric citrate, Ferric Ammonium Citrate, Ammonium iron (III) citrate

09713-1KG 09713-500G 09713-50G
09713-1KGEA 1100 NOK
09713-1KG 09713-500G 09713-50G
Ammoniumjern(III)citrat (14,5 - 16,0% Fe) (RT), green, powder, Purum p.a.
Ammoniumjern(III)citrat

  • Products with a guarantee certificate and/or suitable for the stated analytical application.

Formel: C₆H₅+4y)Fe(x)N(y)O₇
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00013099
CAS-nummer: 1185-57-5
EINECS: 214-686-6
Merck Indeks: 13,00518

Bestill nå

Spesifikasjon Testresultater

Assay of Fe 14.5 - 16.0 %
Calcium (Ca) Max. 1000 ppm
Cadmium (Cd) Max. 50 ppm
Cobalt (Co) Max. 50 ppm
Copper (Cu) Max. 50 ppm
Potassium (K) Max. 500 ppm
Sodium (Na) Max. 5000 ppm
Nickel (Ni) Max. 100 ppm
Lead (Pb) Max. 50 ppm
Zinc (Zn) Max. 50 ppm
Chloride (Cl) Max. 500 ppm
Sulfate (SO₄) Max. 500 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR