Aceton-D6 Isotope_(99.9% D), MagniSolv™ NMR, Sigma-Aldrich®

Leverandør: Merck
Danger

Synonymer: Propanone-D6, DMK-D6, Hexadeuteroacetone, Dimethyl ketone-D6, Acetone-[²H₆], Acetone-d6

MQ200

1.00021.0100
1.00021.0100EA 7250 NOK
1.00021.0100
Aceton-D6 Isotope_(99.9% D), MagniSolv™ NMR, Sigma-Aldrich®
Aceton-D6
Formel: D₃CC(O)CD₃
MW: 64.03 g/mol
Kokepunkt: 55.5
Smeltepunkt: –95.4 °C
Tetthet: 0.79 g/cm³ (20 °C)
Flash punkt: –19 °C (closed cup)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00044635
CAS-nummer: 666-52-4
EINECS: 211-563-9
FN: 1090
ADR: 3 II

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR