Testsett, Klortest for Spectroquant®, Supelco®
Leverandør: Merck
1.00597.0001 1.00598.0001 1.00598.0002 1.00599.0001 1.00602.0001 1.00602.0002 1.00595.0001 1.00086.0001 1.00087.0001 1.00088.0001
1.00597.0001EA 1190 NOK
1.00597.0001 1.00598.0001 1.00598.0002 1.00599.0001 1.00602.0001 1.00602.0002 1.00595.0001 1.00086.0001 1.00087.0001 1.00088.0001
Testsett, Klortest for Spectroquant®, Supelco®
Miljø testkit
All Spectroquant® test kits can be used with the Prove range of spectrophotometers and Nova 60/60A instruments. Tests can be used not only on photometers and spectrophotometer from Merck, but also on photometers and spectrophotometers from other suppliers (programming details available on request). Spectroquant® cell test kits.
  • Very user friendly, validated methods
  • All the necessary reagents for the analysis are contained in the cells for most the test parameters
  • Packaging design protects the cells from fluctuation of temperature and moisture during storage and transport
  • The barcode on the cells ensures the correct method of analysis is chosen - eliminating mistakes and reducing set-up times
Spectroquant® reagent test kits
- Highly stable, ready to use reagent mixtures in a user friendly design
- Free selection of appropriate cell format according to the measuring range
- Long shelf life up to 3 years - all at room temperature
- Automatic selection of the correct method with help of AutoSelector barcode identification
Note: The range quoted in the table below refers to the Prove and NOVA instruments. For some test kits the measuring range available for the Spectroquant® Multy and Move 100 instruments may be reduced.
*4 Reagenser for testing av fritt klor er reagensen Cl₂-1 1.00086.0001 og reagensen Cl₂-2 1.00087.0001 sammen med tomme kyvetter og tilbehør 1.00089.0001
Reagenser for total klor er reagensen Cl₂-1 1.00086.0001, reagensen Cl₂-2 1.00087.0001 og reagensen Cl₂-3 1.00088.0001 sammen med tomme kyvetter og tilbehør 1.00089.0001
*7 Denne metoden er offisielt godkjent av USEPA som en alternativ metode for å undersøke følgende:
*7-1 avløpsvann
*7-2 drikkevann
*7-3 henholdsvis drikkevann og avløpsvann
Bestill nå
SPESIFIKASJONER
Environmentally Preferable
M-Clarity quality level MQ100