Nuclear fast red-aluminum sulphate solution 0,1% used in nuclear staining, for histologi, Sigma-Aldrich®
Leverandør: Merck
Danger
1.00121.0500
1.00121.0500EA 845 NOK
1.00121.0500
Nuclear fast red-aluminum sulphate solution 0,1% used in nuclear staining, for histologi, Sigma-Aldrich®
Nuclear fast red-aluminum sulphate solution
For nuclear staining and for counterstaining in histochemical reactions e.g. Berlin blue
Nuclear fast red-aluminum sulfate solution 0.1% is used for counterstaining in histochemical reactions, e.g. in Berlin blue staining for detection of iron or in silver staining of reticulin fibers. Nuclear fast red-aluminum sulfate solution 0.1% can also be used for nuclear staining with Alcian blue. 500 ml of the Nuclear fast red-aluminum sulfate solution 0.1% are sufficient for staining at least 500 specimens. The solution has an excellent quality as it is an IVD registered product and is CE certified, thus, it can be used in an accredited laboratory.
IVDLogo
Bestill nå
Spesifikasjon Testresultater
Suitability for microscopy passes test
Nuclei dark red
Cytoplasm light red
Acid mucosubstances light blue