Benzosyre, EMSURE® Reag. Ph. Eur. for analysis, Supelco®

Leverandør: Merck
Danger


M-Clarity™ quality level = MQ300

1.00136.0100 1.00136.0250 1.00136.1000 1.00136.9025
1.00136.0100EA 545 NOK
1.00136.0100 1.00136.0250 1.00136.1000 1.00136.9025
Benzosyre, EMSURE® Reag. Ph. Eur. for analysis, Supelco®
Benzosyre
Formel: C₇H₆O₂
Kokepunkt: 249 °C (1013 hPa)
Smeltepunkt: 121…123 °C
Tetthet: 1,316 g/cm³ (20 °C)
Flash punkt: 121 °C (closed cup)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00002398
CAS-nummer: 65-85-0
EINECS: 200-618-2
REACH: 01-2119455536-33
Merck Indeks: 13,01092
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d561f311b11ea8ab2c8f07253e6748e588eb71aa&vid=dcf5e45072cda5106216b497cd29716270ae772c&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Solubility 2.9 g/l (25 °C)
Melting Point 121 - 123 °C
Molar Mass 122.12 g/mol
Bulk Density 500 kg/m3
Boiling Point 249 °C (1013 hPa)
Vapor Pressure 0.001 hPa (20 °C)
Flash Point 121 °C
Density 1.321 g/cm3 (20 °C)
pH 2.5 - 3.5 (H2O, 20 °C) (saturated solution)
Ignition Point 570 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR