Chromocult® RAMBACH™ Agar ISO 6579 for Selective Isolation of Salmonella spp., Millipore
Leverandør: Merck
1.00188.0002 1.00188.0004
1.00188.0002EA 1550 NOK
1.00188.0002 1.00188.0004
Chromocult® RAMBACH™ Agar ISO 6579 for Selective Isolation of Salmonella spp., Millipore
Mikrobiologiske medier Media, dehydrert for mikrobiologi
For the isolation and differentiation of Salmonella from food and animal feed, water and other materials. RAMBACH® agar ref. to ISO 6579 is also known as Propylene glycol deoxycholate neutral red agar.
  • For the identification of Salmonella
  • Complies to EN ISO 6579-1
Analysis Note
The dehydrated medium appears as dark-green color.
The prepared medium is opaque and light pink in appearance.
The pH value at 25 degrees is in the range of 7.1-7.5.
Sertifiseringer: This culture medium complies with the specifications reference to EN ISO 6579-1.
This culture medium is released by the quality control laboratory of Merck KGaA, Darmstadt, Germany. The laboratory is accredited by the German accreditation authority DAkkS as registered test laboratory D-PL-15185-01-00 according to DIN EN ISO/IEC 17025 for the performance testing of media for microbiology according to DIN EN ISO 11133.
CHROMOCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
Rambach is a trademark of Balel S.A.
Bestill nå
SPESIFIKASJONER
Viktige bruksområder Selective for enterobacteriaceae: Salmonella spp.
Environmentally Preferable
Applikasjon Microbiological culture
Skjema Powder
pH 7.3±0.2 (25 °C)
Lagringstemp 15-25°C
M-Clarity quality level MQ200