MacCONKEY Agar acc. ISO 21150, ISO 21567, FDA-BAM and EP, USP, JP GranuCult® prime, Millipore
Leverandør: Merck
1.00205.0500 1.00205.5000
1.00205.0500EA 1410 NOK
1.00205.0500 1.00205.5000
MacCONKEY Agar acc. ISO 21150, ISO 21567, FDA-BAM and EP, USP, JP GranuCult® prime, Millipore
Mikrobiologiske medier Media, dehydrert for mikrobiologi
For the isolation of Salmonella, Shigella and coliform bacteria from foodstuffs and wastewater.