Sitronsyre monohydrat, EMSURE® ACS, ISO, Reag. Ph. Eur. for analysis, Supelco®

Leverandør: Merck
Warning


M-Clarity™ quality level = MQ300

1.00244.0500 1.00244.1000 1.00244.5000 1.00244.9026
1.00244.0500EA 767 NOK
1.00244.0500 1.00244.1000 1.00244.5000 1.00244.9026
Sitronsyre monohydrat, EMSURE® ACS, ISO, Reag. Ph. Eur. for analysis, Supelco®
Sitronsyre monohydrat

Sertifiseringer: Corresponds to ACS, ISO 6353/2, Reag. Ph. Eur.

Formel: C₆H₈O₇.H₂O
Smeltepunkt: 135…152 °C
Tetthet: 1,552 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00149972
CAS-nummer: 5949-29-1
EINECS: 201-069-1
Merck Indeks: 13,02350
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3160681f96abd248f56672a96b72cc5f6e9d89a2&vid=c4db52c32c630d2c9876e60fc3e9e70a117ea0d8&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Assay (acidimetric, calc. on anhydrous substance) 99.5 - 100.5 %
Identity (IR-spectrum) passes test
Appearance white to almost white or colorless crystals
Appearance of solution (200 g/l; water) clear (≤ 3 NTU) and not more intense in color than reference solution Y₇, BY₇ or GY₇
In water insoluble matter ≤ 50 ppm
Chloride (Cl) ≤ 5 ppm
Phosphate (PO₄) ≤ 10 ppm
Sulphate (SO₄) ≤ 20 ppm
Heavy metals (as Pb) ≤ 5 ppm
Al (Aluminium) ≤ 0.2 ppm
As (Arsenic) ≤ 1 ppm
Hg (Mercury) ≤ 1 ppm
Cu (Copper) ≤ 5 ppm
Fe (Iron) ≤ 3 ppm
Fe (Iron, acc. to Reag. Ph  Eur.) suitable to use in the limit test for iron
Pb (Lead) ≤ 0.5 ppm
Oxalates (as C₂H₂O₄) ≤ 50 ppm
Readily carbonisable substance passes test
Sulfated ash ≤ 200 ppm
Water (according to Karl Fischer) 7.5 - 8.8 %

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR