Colour Reference solution, CertiPUR®, Supelco®

Leverandør: Merck

1.00246.0250
1.00246.0250EA 2740 NOK
1.00246.0250
Colour Reference solution, CertiPUR®, Supelco®
Standarder Fargereferanse og spekroskopistandarer

The colour reference solution can be used in visual comparison or as part of a calibration of colorimeters and spectrophotometer for colour comparison. The solution can be used to prepare reference solution with different Hazen values.


  • Traceable to NIST SRM930e
  • Shelf life of unopened bottle is minimum of 18 months


For use under DIN EN ISO 7887, ASTM D 1209-05, DIN EN ISO 6271-1, APHA 2120 B & C

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR