Reticulum stain kit acc. to Gordon & Sweets, Sigma-Aldrich®

Leverandør: Merck
Danger

1.00251.0001
1.00251.0001EA 4570 NOK
1.00251.0001
Reticulum stain kit acc. to Gordon & Sweets, Sigma-Aldrich®
Reticulum Stain Kit

This reticulin silver plating stain has been designed for the detection of reticular fibres in histological specimens.


Fibres that cannot be adequately visualised using an H and E stain may be visible through this silver plating stain.


Used to visualise human reticular fibres, which are components of membrane structures, capillaries and peripheral nerve fibres, as well as organs such as the liver, kidneys and hematopoietic organs.


Reticulin visualisation gives greater contrast and sharpness against a background.


Gordon and Sweets' silver stain for reticulin may be followed by nuclear staining with nuclear fast red aluminium sulphate solution 0,1% when nuclei, collagen and the background are to be stained in addition to the reticular fibres. Sufficient for up to 110 section slides.


The kit is sufficient for up to 110 section slides.


Forpakning: Eight bottles of 30 ml, in a fibre case.

FN: 3316
ADR: 9 II
IVDLogo

Bestill nå

Spesifikasjon Testresultater

Suitability for microscopy Passes test
Reticular fibres Black
Nuclei Red
Collagen Red
Background Red

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR