Barium difenylamin-4-sulfonat, Millipore®

Leverandør: Merck
Warning


M-Clarity™ quality level = MQ200

1.00255.0005
1.00255.0005EA 1660 NOK
1.00255.0005
Barium difenylamin-4-sulfonat, Millipore®
Barium difenylamin-4-sulfonat
Formel: C₂₄H₂₀BaN₂O₆S₂
Smeltepunkt: 300 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00007497
CAS-nummer: 6211-24-1
EINECS: 228-278-0
FN: 1564
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8c37e55aa4075d4ea7537fbef23f9195faa03f14&vid=5a934374d2d4e697c6ecd2cd9828f06692feca01&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Solubility (20 °C) soluble
Melting Point 300 °C
Molar Mass 633.88 g/mol
Bulk Density 220 kg/m3
pH 5.9 (10 g/l, H2O, 20 °C) (slurry)
Hazard Class 6.1
Hazard Class Description Toxic Materials


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR