Hydrogenbromid 47%, EMSURE® ISO for analysis, Supelco®

Leverandør: Merck
Danger


M-Clarity™ quality level = MQ300

1.00307.1000
1.00307.1000EA 3100 NOK
1.00307.1000
Hydrogenbromid 47%, EMSURE® ISO for analysis, Supelco®
Hydrobromic acid
Formel: HBr
Tetthet: 1,47 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00011323
CAS-nummer: 10035-10-6
EINECS: 233-113-0
FN: 1788
ADR: 8,II

Bestill nå

Spesifikasjon Testresultater

Solubility (20 °C) soluble
Melting Point -11 °C
Boiling Point 126 °C (1013 hPa)
Vapor Pressure 10.6 hPa (20 °C)
Density 1.49 g/cm3 (20 °C)
pH (H2O, 20 °C) strongly acid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR