N,N-Dimetylacetamid, SupraSolv® for headspace gas chromatography, Supelco®

Leverandør: Merck
Danger

Synonymer: Acetic acid dimethylamide, Dimethyl Acetamide, DMAC

M-Clarity™ quality level = MQ100

1.00399.1000
1.00399.1000EA 2620 NOK
1.00399.1000
N,N-Dimetylacetamid, SupraSolv® for headspace gas chromatography, Supelco®
N,N-Dimetylacetamid
Formel: CH₃CON(CH₃)₂
MW: 87.12 g/mol
Kokepunkt: 166.1
Smeltepunkt: –20 °C
Tetthet: 0.937 g/cm³ (20 °C)
Flash punkt: 70
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00008686
CAS-nummer: 127-19-5
EINECS: 204-826-4
FN: 2810
Merck Indeks: 13,03254
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ace04c31b8db2d1fc0c18051cd2eefc52a61ea65&vid=4e98bd4c34a9326a56ee3e11797b2e6e1b323c8f&prid=web&clid=SAPDAM

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR