HPLC columns, Hibar®, LiChrospher® 100 NH₂
Leverandør: Merck
1.00423.7076 1.00424.7076 1.00424.7077 1.50009.7076 1.50009.7077 1.50012.7076 1.50012.7077 1.50013.7076 1.50013.7077 1.50182.7076 1.50182.7077 1.50183.7076 1.50183.7077
1.00423.7076EA 12200 NOK
1.00423.7076 1.00424.7076 1.00424.7077 1.50009.7076 1.50009.7077 1.50012.7076 1.50012.7077 1.50013.7076 1.50013.7077 1.50182.7076 1.50182.7077 1.50183.7076 1.50183.7077
HPLC columns, Hibar®, LiChrospher® 100 NH₂
Kromatografikolonner
Stainless steel Hibar® columns are complete with endfittings. These columns are available with various types of LiChrospher® sorbents.
When using a guard column with a Hibar® RT column, we recommend part number 1.51487.0001 guard column cartridge holder for 4×4 mm guard column cartridges LiChroCART®.