HPLC columns, Hibar®, with Superspher® Sorbents
Leverandør: Merck
1.00423.7137 1.00423.7138 1.00423.7139 1.00423.7140 1.00423.7141 1.00423.7142 1.00424.7137 1.00424.7138 1.00424.7139 1.00424.7140 1.00424.7142 1.50009.7137 1.50009.7138 1.50009.7139 1.50009.7140 1.50009.7141 1.50009.7142 1.50012.7137 1.50012.7138 1.50012.7139 1.50012.7140 1.50012.7141 1.50012.7142 1.50013.7137 1.50013.7138 1.50013.7139 1.50013.7140 1.50013.7141 1.50013.7142 1.50182.7138 1.50182.7140 1.50182.7142 1.50183.7137 1.50183.7138 1.50183.7139 1.50183.7140 1.50183.7141 1.50183.7142 1.00423.7143
1.00423.7137EA 12200 NOK
1.00423.7137 1.00423.7138 1.00423.7139 1.00423.7140 1.00423.7141 1.00423.7142 1.00424.7137 1.00424.7138 1.00424.7139 1.00424.7140 1.00424.7142 1.50009.7137 1.50009.7138 1.50009.7139 1.50009.7140 1.50009.7141 1.50009.7142 1.50012.7137 1.50012.7138 1.50012.7139 1.50012.7140 1.50012.7141 1.50012.7142 1.50013.7137 1.50013.7138 1.50013.7139 1.50013.7140 1.50013.7141 1.50013.7142 1.50182.7138 1.50182.7140 1.50182.7142 1.50183.7137 1.50183.7138 1.50183.7139 1.50183.7140 1.50183.7141 1.50183.7142 1.00423.7143
HPLC columns, Hibar®, with Superspher® Sorbents
Kromatografikolonner
Stainless steel Hibar® columns are complete with endfittings. These columns are available with various types of Superspher® sorbents.
When using a guard column with a Hibar® RT column, we recommend part number 1.51487.0001 guard column cartridge holder for 4×4 mm guard column cartridges LiChroCART®.