Salpetersyre 65%, EMSURE® ISO for analysis (maks. 0,0000005% Hg), Supelco®

Leverandør: Merck
Danger


M-Clarity™ quality level = MQ300

1.00452.1000 1.00452.2500 1.00452.9180
1.00452.1000EA 492 NOK
1.00452.1000 1.00452.2500 1.00452.9180
Salpetersyre 65%, EMSURE® ISO for analysis (maks. 0,0000005% Hg), Supelco®
Salpetersyre
Formel: HNO₃
Kokepunkt: 120,5 °C (1013 hPa)
Smeltepunkt: –42 °C
Tetthet: 1,4 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00011349
CAS-nummer: 7697-37-2
EINECS: 231-714-2
FN: 2031
ADR: 8 (5.1),II
Merck Indeks: 13,06608

Bestill nå

Spesifikasjon Testresultater

Solubility (20 °C) soluble
Melting Point -32 °C
Boiling Point 121 °C (1013 hPa)
Vapor Pressure 9.4 hPa (20 °C)
Density 1.39 g/cm³ (20 °C)
pH (H₂O, 20 °C) strongly acid
Hazard Class 8
Hazard Class Description Corrosive material

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR