Vann, Tracepur® for inorganic trace analysis, Supelco®

Leverandør: Merck

Synonymer: Hydrogen oxide

M-Clarity™ quality level = MQ100

1.00473.5000
1.00473.5000EA 6320 NOK
1.00473.5000
Vann, Tracepur® for inorganic trace analysis, Supelco®
Vann
Formel: H₂O
Kokepunkt: 100 °C (1013 hPa)
Smeltepunkt: 0 °C
Tetthet: 1 g/cm³ (4 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00011332
CAS-nummer: 7732-18-5
EINECS: 231-791-2
Merck Indeks: 13,10098

Bestill nå

Spesifikasjon Testresultater

Molar Mass 18.02 g/mol
Boiling Point 100 °C (1013 hPa)
Vapor Pressure 23 hPa (20 °C)
Density 1.00 g/cm3 (20 °C)
pH (H2O, 20 °C) neutral


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR