Dodecamolybdophosphoric acid solution_SC 2-propanol, spray_reagent TLC, Supelco®

Leverandør: Merck
Danger

Synonymer: Phosphomolybdic acid, Molybdophosphoric acid

MQ100

1.00480.0100
1.00480.0100EA 743 NOK
1.00480.0100
Dodecamolybdophosphoric acid solution_SC 2-propanol, spray_reagent TLC, Supelco®
Dodecamolybdophosphoric acid

Ready-to-use spray solution.

Formel: H₃Mo₁₂O₄₀P
MW: 1825.25 g/mol
MDL nummer: MFCD00011341
CAS-nummer: 12026-57-2
FN: 1219
ADR: 3 II

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR