Oksalsyre (Etandisyre) dihydrat, Suprapur® for trace analysis, Supelco®

Leverandør: Merck
Danger


M-Clarity™ quality level = MQ100

1.00489.0100
1.00489.0100EA 3775 NOK
1.00489.0100
Oksalsyre (Etandisyre) dihydrat, Suprapur® for trace analysis, Supelco®
Oksalsyre (Etandisyre) dihydrat
Formel: HO₂CCO₂H·2H₂O
Kokepunkt: 149…160 °C (1013 hPa)
Smeltepunkt: 101 °C
Tetthet: 1,65 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00149102
CAS-nummer: 6153-56-6
EINECS: 205-634-3
FN: 3261
ADR: 8,III
REACH: 01-2119534576-33
Merck Indeks: 13,06980
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dcc1447f5f23a589dc85e10dd090c86e33b6541b&vid=2f0e34543b2f625715b002905fc0dd45fcd75561&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Solubility 102 g/l (20 °C)
Melting Point 101 °C
Molar Mass 126.07 g/mol
Bulk Density 900 kg/m3
Boiling Point 149 - 160 °C (decomposition)
Vapor Pressure 21 hPa (50 °C)
Flash Point 157 °C (decomposition)
Density 1.65 g/cm3 (20 °C)
pH 1 (10 g/l, H2O, 20 °C)
Hazard Class 8
Hazard Class Description Corrosive Material


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR