Perchloric acid 70% ≥70%, Suprapur® for trace analysis, Supelco®

Leverandør: Merck
Danger


M-Clarity™ quality level = MQ100

1.00517.0250 1.00517.1000
1.00517.0250EA 4470 NOK
1.00517.0250 1.00517.1000
Perchloric acid 70% ≥70%, Suprapur® for trace analysis, Supelco®
Perchloric acid
Formel: HClO₄
Kokepunkt: 160…181 °C (1013 hPa)
Tetthet: 1,53…1.768 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00011325
CAS-nummer: 7601-90-3
EINECS: 231-512-4
FN: 1873
ADR: 5.1 (8),I
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5fc15d8358229daccc96a56573b32589d2a21575&vid=3780c3029836d8b95181cdef71dc2669f7e2a797&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Solubility (20 °C) soluble
Melting Point -18 °C
Boiling Point 198.7 °C (1013 hPa)
Density 1.68 g/cm3 (20 °C)
pH (H2O, 20 °C) strongly acid
Hazard Class 5.1
Hazard Class Description Oxidizer


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR