Dodecamolybdofosforsyre hydrat, EMSURE® ACS, Reag. Ph. Eur. for analysis, Supelco®

Leverandør: Merck
Danger


M-Clarity™ quality level = MQ300

1.00532.0025 1.00532.0100
1.00532.0025EA 1280 NOK
1.00532.0025 1.00532.0100
Dodecamolybdofosforsyre hydrat, EMSURE® ACS, Reag. Ph. Eur. for analysis, Supelco®
Dodecamolybdofosforsyre hydrat
Formel: H₃[P(Mo₃O₁₀)₄]·nH₂O
Smeltepunkt: 78…90 °C
Tetthet: 1,62 g/cm³ (25 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00149913
CAS-nummer: 51429-74-4
EINECS: 234-713-5
FN: 3260
ADR: 8,II
Merck Indeks: 12,07498

Bestill nå

Spesifikasjon Testresultater

Solubility (20 °C) soluble
Melting Point 78 - 90 °C (anhydrous substance)
Molar Mass 1825.28 g/mol (anhydrous substance)
Bulk Density 980 kg/m3
pH (H2O) strongly acid

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR