Granular dessicant, Sicapent® with indicator, Supelco®
Leverandør: Merck
1.00543.0500
1.00543.0500EA 980 NOK
1.00543.0500
Granular dessicant, Sicapent® with indicator, Supelco®
Eksikatorer Tørkemidler
Sicapent is a granular dessicant comprising ~75% phoshorus pentoxide on an inert inorganic carrier.
  • Ease of use
  • Fast water adsorption due to large surface area
  • Colour indicator - colourless to green to blue
  • Remains in particle form after 100% water uptake
Appliations include drying liquids, filling drying tubes, particularly for fast flowing gases in drying tubes.
Bulk density: ~300 g/L