Salisylsyre, fint pulver, EMPROVE® ESSENTIAL Ph. Eur., BP, USP, SAFC®

Leverandør: MERCK PRODUCTION CHEMICALS
Danger

1.00635.9019
1.00635.9019EA 40650 NOK
1.00635.9019
Salisylsyre, fint pulver, EMPROVE® ESSENTIAL Ph. Eur., BP, USP, SAFC®
Salisylsyre
Formel: (HO)C₆H₄CO₂H
Kokepunkt: 336 °C (1013 hPa)
Smeltepunkt: 157…159 °C
Tetthet: 1,443 g/cm³ (20 °C)
Flash punkt: 157 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00002439
CAS-nummer: 69-72-7
EINECS: 200-712-3
Merck Indeks: 13,08411
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=af1f695fa92656a135b5e1742f1cca28f4d255fb&vid=8f3aff6fb9a70e5dd194b8bc46019af74e48e90a&prid=web&clid=SAPDAM

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR