Svovelsyre 95-97%, EMSURE® ACS, ISO, Reag. Ph. Eur. for analysis (maks. 0,0000005% Hg), Supelco®

Leverandør: Merck
Danger


M-Clarity™ quality level = MQ300

1.00732.2500 1.00732.1000 1.00732.2510 1.00732.0510
1.00732.2500EA 943.55 NOK
1.00732.2500 1.00732.1000 1.00732.2510 1.00732.0510
Svovelsyre 95-97%, EMSURE® ACS, ISO, Reag. Ph. Eur. for analysis (maks. 0,0000005% Hg), Supelco®
Svovelsyre
Formel: H₂SO₄
Kokepunkt: ∼335 °C (1013 hPa)
Smeltepunkt: 3 °C
Tetthet: 1,84 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00064589
CAS-nummer: 7664-93-9
EINECS: 231-639-5
FN: 1830
ADR: 8,II
Merck Indeks: 14,08974

Bestill nå

Spesifikasjon Testresultater

Solubility (20 °C) soluble,(caution ! development of heat)
Melting Point -20 °C
Molar Mass 98.08 g/mol
Boiling Point 335 °C
Vapor Pressure 0.0001 hPa (20 °C)
Density 1.84 g/cm3 (20 °C)
pH 0.3 (49 g/l, H2O, 25 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR