Etylacetat ≥99.8%, SupraSolv® for GC-MS

Leverandør: Merck
Danger


M-Clarity™ quality level = MQ100

1.00789.1000 1.00789.2500
1.00789.1000EA 825 NOK
1.00789.1000 1.00789.2500
Etylacetat ≥99.8%, SupraSolv® for GC-MS
Etylacetat
Formel: CH₃COOC₂H₅
Kokepunkt: 77,1 °C (1013 hPa)
Smeltepunkt: –83 °C
Tetthet: 0,902 g/cm³ (20 °C)
Flash punkt: –4 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00009171
CAS-nummer: 141-78-6
EINECS: 205-500-4
FN: 1173
ADR: 3,II
REACH: 01-2119475103-46
Merck Indeks: 13,03792
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=226aecffe42ffab125597552eff0ef7f88ef9983&vid=f5707ce0faee09ca10700419252c5841e0fbc01a&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Purity (GC) =99.8%
Identity(IR) Conforms
Evaporation Residue = 3.0 mg/l
Water = 0.02%
Color =10 Hazen
GC/MSD (retention range n-undecane to n-tetracontane, scanning area 30-600 amu, individual signals (n-tetradecane standard)) = 3ng/ml
Suitability Residue Analysis


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR