Dietyleter, Uvasol® for spectroscopy, Supelco®

Leverandør: Merck
Danger

Synonymer: Ethyl Ether, Ether, 2,2'-Oxydiethane, 2,2'-Oxybisethane, Ethoxyethane

M-Clarity™ quality level = MQ100

1.00930.1000
1.00930.1000EA 1890 NOK
1.00930.1000
Dietyleter, Uvasol® for spectroscopy, Supelco®
Dietyleter
Formel: (CH₃CH₂)₂O
MW: 74.12 g/mol
Kokepunkt: 35
Smeltepunkt: –123 °C
Tetthet: 0.7135 g/cm³ (20 °C)
Flash punkt: –40 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00011646
CAS-nummer: 60-29-7
EINECS: 200-467-2
FN: 1155
ADR: 3 I
REACH: 01-2119535785-29
Merck Indeks: 12,03852
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=16f2debd1f18f9bff9952c4a6e5d599a67f94e61&vid=3dd6c545e58e36e1004e828026523fb0e3bcbbcd&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Solubility 69 g/L (20 °C)
Melting Point -116.3 °C
Molar Mass 74.12 g/mol
Boiling Point 34.6 °C (1013 hPa)
Vapor Pressure 587 hPa (20 °C)
Flash Point -40 °C
Explosion Limit 1.7 - 36%(V)
Density 0.71 g/cm³
Ignition Point 180 °C
Saturation Concentration 1776 g/m³ (20 °C) Air
Evaporation Number 1
Hazard Class 3
Hazard Class Description Flammable liquid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR