Isobutanol, EMPLURA®, Supelco®

Leverandør: Merck
Danger


M-Clarity™ quality level = MQ200

1.00985.6025 1.00985.9190
1.00985.6025EA 8310 NOK
1.00985.6025 1.00985.9190
Isobutanol, EMPLURA®, Supelco®
Isobutanol
Formel: (CH₃)₂CHCH₂OH
Kokepunkt: 107,9 °C (1013 hPa)
Smeltepunkt: –108 °C
Tetthet: 0,802 g/cm³ (20 °C)
Flash punkt: 28 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00004740
CAS-nummer: 78-83-1
EINECS: 201-148-0
FN: 1212
ADR: 3,III
Merck Indeks: 13,05148
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=de7c83a107c7a71fa54ed2b1bbdfb78617fe3f67&vid=df3ce90fc7ae8dafb7fabe31b244825735325030&prid=web&clid=SAPDAM

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR