2-Propanol, SupraSolv® for gasskromatografi, Supelco®

Leverandør: Merck
Danger


M-Clarity™ quality level = MQ100

1.00998.1000 1.00998.2500
1.00998.1000EA 575 NOK
1.00998.1000 1.00998.2500
2-Propanol, SupraSolv® for gasskromatografi, Supelco®
2-Propanol
Formel: (CH₃)₂CHOH
Kokepunkt: 82 °C (1013 hPa)
Smeltepunkt: –89 °C
Tetthet: 0,786 g/cm³ (20 °C)
Flash punkt: 12 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00011674
CAS-nummer: 67-63-0
EINECS: 200-661-7
FN: 1219
ADR: 3,II
REACH: 01-2119457558-25
Merck Indeks: 13,05228
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ceac541820c6046c770c320cb35e80c1343901a5&vid=4a720b44a08c22b968be82ae24c2b3d604bdd52b&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Solubility (20 °C) soluble
Melting Point -89.5 °C
Molar Mass 60.1 g/mol
Boiling Point 82.4 °C (1013 hPa)
Vapor Pressure 43 hPa (20 °C)
Flash Point 12 °C
Refractive Index 1.378
Explosion Limit 2 - 13.4 %(V)
Density 0.786 g/cm3 (20 °C)
pH (H2O, 20 °C) neutral
Ignition Point 425 °C
Evaporation number 11
Water absorption 1000 g/kg
Hazard Class 3
Hazard Class Description Flammable liquid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR