Ammonium carbamate, Supelco®

Leverandør: Merck
Danger

Synonymer: Carbamic acid ammonium salt

M-Clarity™ quality level = MQ300

1.01134.0500
1.01134.0500EA 1460 NOK
1.01134.0500
Ammonium carbamate, Supelco®
Ammonium carbamate
Formel: H₂NCO₂NH₄
MW: 78.07 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00013010
CAS-nummer: 1111-78-0
EINECS: 214-185-2

Bestill nå

Spesifikasjon Testresultater

Solubility 790 g/l (20 °C)
Melting Point 152 °C (closed capillary tube)
Molar Mass 78.07 g/mol
Bulk Density 750 kg/m3
Vapor Pressure 82 hPa (20 °C)
Density 1.6 g/cm3 (20 °C)
pH 10 (100 g/l, H2O, 20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR