Ammoniumsulfat, Millipore®

Leverandør: Merck


M-Clarity™ quality level = MQ100

1.01211.1000 1.01211.5000 1.01211.9050
1.01211.1000EA 617 NOK
1.01211.1000 1.01211.5000 1.01211.9050
Ammoniumsulfat, Millipore®
Ammonium sulphate
Formel: (NH₄)₂SO₄
Smeltepunkt: 336…339 °C
Tetthet: 1,78 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00003391
CAS-nummer: 7783-20-2
EINECS: 231-984-1
REACH: 01-2119455044-46
Merck Indeks: 13,00559

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR