Anilin, EMSURE® for analysis, Supelco®

Leverandør: Merck
Danger

Synonymer: Aminobenzene, Phenylamine

M-Clarity™ quality level = MQ300

1.01261.1000
1.01261.1000EA 1270 NOK
1.01261.1000
Anilin, EMSURE® for analysis, Supelco®
Anilin
Formel: C₆H₅NH₂
MW: 93.13 g/mol
Kokepunkt: 184
Smeltepunkt: –6.2 °C
Tetthet: 1.019 g/cm³ (20 °C)
Flash punkt: 76
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00007629
CAS-nummer: 62-53-3
EINECS: 200-539-3
FN: 1547
ADR: 6.1 II
Merck Indeks: 13,00661
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3fc5b5071f5a578057c8a7c1c4855ca964c4d840&vid=4f9c118a47c84e5b067d971f00a4eee6dc846d80&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Solubility 36 g/l (20 °C)
Melting Point -6.2 °C
Molar Mass 93.13 g/mol
Boiling Point 184 °C (1013 hPa)
Vapor Pressure 0.5 hPa (20 °C)
Flash Point 76 °C
Refractive Index 1.5863 (20 °C, 589 nm)
Explosion Limit 1.2 - 11 %(V)
Density 1.02 g/cm3 (20 °C)
pH 8.8 (36 g/l, H2O, 20 °C)
Ignition Point 540 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR