Acid green 1 indicator (C.I. 10020), Millipore®

Leverandør: Merck

Synonymer: Naphthol Green B, Iron, tris(5,6-dihydro-5,6-dioxo-2-naphthalenesulphonic acid 5-oximato)-,trisodium salt

M-Clarity™ quality level = MQ200

1.01306.0025
1.01306.0025EA 758 NOK
1.01306.0025
Acid green 1 indicator (C.I. 10020), Millipore®
Acid green 1
Formel: C₃₀H₁₈FeN₃O₁₅S₃
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00003886
CAS-nummer: 19381-50-1
EINECS: 243-010-2

Bestill nå

Spesifikasjon Testresultater

Solubility 160 g/l
Molar Mass 878.46 g/mol
Bulk Density 620 kg/m3
pH 9.3 (10 g/l, H2O, 20 °C)
Bulk density 620 kg/m3
Storage temperature +5...+30 °C

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR