Wright's stain solution

Leverandør: Merck
Danger

Synonymer: Eosin Methylene blue solution according to Wright, Wright's eosin methylene blue

1.01383.0500 1.01383.2500 1.01383.1022 1.01383.9180 1.01383.9025
1.01383.0500EA 320 NOK
1.01383.0500 1.01383.2500 1.01383.1022 1.01383.9180 1.01383.9025
Wright's stain solution
Wright's stain

Used to stain blood, bone marrow smears, and clinical cytological specimens


The staining method according to Wright is one of the standard techniques in hematological diagnostic procedures. Like Leishman's eosin methylene blue solution and May-Grünwald's eosin methylene blue solution, it is used for staining of blood and bone marrow smears and clinical cytological specimens. The Wright’s eosin methylene blue solution is an IVD / CE product and 500 ml are sufficient for staining up to 1600 slides in Hellendahl cells.

Tetthet: 0.80 g/cm3 (20 °C)
Flash punkt: 9.7 °C
MDL nummer: MFCD00082143
CAS-nummer: 68988-92-1
FN: 1230
ADR: 3 (6.1) II
IVDLogo

Bestill nå

Spesifikasjon Testresultater

Suitability for microscopy (Blood smear) passes test
Erythrocytes pink to brownish
Nuclei blue-violet
Eosinophilic granules red to red-brown
Neutrophilic granules light violet
Lymphocyte cytoplasm Blue


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR