May Grunwald's eosin-methylene blue modified solution stain for hematology, for cytology, Sigma-Aldrich®

Leverandør: Merck
Danger


M-Clarity™ quality level = MQ300

1.01424.0100 1.01424.0500 1.01424.2500 1.01424.9025 1.01424.1000
1.01424.0100EA 283 NOK
1.01424.0100 1.01424.0500 1.01424.2500 1.01424.9025 1.01424.1000
May Grunwald's eosin-methylene blue modified solution stain for hematology, for cytology, Sigma-Aldrich®
May Grunwald's eosin-methylene blue

Used to stain blood, bone marrow smears, and clinical cytological specimens

Tetthet: 0.79 g/cm³ (20 °C)
MDL nummer: MFCD00131580
FN: 1230
ADR: 3 (6.1) II
IVDLogo

Bestill nå

Spesifikasjon Testresultater

Suitability for microscopy (Blood smear) passes test
Erythrocytes pink to brownish
Nuclei violet
Eosinophilic granules red-brown
Neutrophilic granules light violet
Lymphocyte cytoplasm Blue


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR