Salpetersyre 60%, Ultrapure for sporanalyse, Supelco®

Leverandør: Merck
Danger


M-Clarity™ quality level = MQ100

1.01518.0250 1.01518.0500 1.01518.1000
1.01518.0250EA 8160 NOK
1.01518.0250 1.01518.0500 1.01518.1000
Salpetersyre 60%, Ultrapure for sporanalyse, Supelco®
Salpetersyre
Formel: HNO₃
Kokepunkt: ∼120 °C (1013 hPa)
Smeltepunkt: –22 °C
Tetthet: 1,37 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00011349
CAS-nummer: 7697-37-2
EINECS: 231-714-2
FN: 2031
ADR: 8,II

Limited Time Sale

Select items below are currently on sale

Bestill nå

Spesifikasjon Testresultater

Solubility (20 °C) soluble
Melting Point -22 °C
Boiling Point 120 °C (1013 hPa)
Density 1.37 g/cm3 (20 °C)
pH  - 1 (H2O, 20 °C)
Hazard Class 8
Hazard Class Description Corrosive Material

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR