1-klorbutan, LiChrosolv® for HPLC, Supelco®

Leverandør: Merck
Danger

Synonymer: n-Butyl Chloride

M-Clarity™ quality level = MQ100

1.01692.1000
1.01692.1000EA 2130 NOK
1.01692.1000
1-klorbutan, LiChrosolv® for HPLC, Supelco®
1-klorbutan
Formel: CH₃(CH₂)₃Cl
MW: 92.57 g/mol
Kokepunkt: 78.4
Smeltepunkt: –123 °C
Tetthet: 0.8862 g/cm³ (20 °C)
Flash punkt: –17 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00001009
CAS-nummer: 109-69-3
EINECS: 203-696-6
FN: 1127
ADR: 3 II
Merck Indeks: 13,01559
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e2bd980eb6e7ecd91951d46d0ea510119e976dc0&vid=5231549172a609ad96623a80204b11c7c0c69273&prid=web&clid=SAPDAM

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR